Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasticoid Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles
SDP

$310.50 - $552.00

Specifications

Color Red
Material Rubber
Product Type Sleeve Stopper
For Use With (Equipment) Serum and vaccine bottles
View More Specs

Products 4
Catalog Number Mfr. No. Size Stopper Number Length (Metric) Sleeve Price Quantity & Availability  
Catalog Number Mfr. No. Size Stopper Number Length (Metric) Sleeve Price Quantity & Availability  
03-215-5
SDP
View Documents
Plasticoid
20200
11.5 11.5 1cm
Pack of 100 for $310.50
Only null left
Add to Cart
 
14-126DD
SDP
View Documents
Plasticoid
20230
15 19 1.9cm
Pack of 100 for $552.00
Only null left
Add to Cart
 
14-126AA
SDP
View Documents
Plasticoid
20210
13 13 1.46cm
Pack of 100 for $355.00
Only null left
Add to Cart
 
14-126BB
SDP
View Documents
Plasticoid
20220
14.5 14.5 1.5cm
Pack of 100 for $477.50
Only null left
Add to Cart
 
Description

Description

  • Comprised of dry natural rubber
  • Folds down over neck of bottle
  • With hollow plugs, rounded corners and needle guide
  • Easily perforated with hypodermic needles
  • Self-sealing

Specifications

Specifications

Red
Rubber
Sleeve Stopper
Serum and vaccine bottles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.