Learn More
Electron Microscopy Sciences Press-To-Seal Red Silicone Sheet, 0.5 mm thick
SDP
Supplier:  Electron Microscopy Sciences 7033870
Encompass_Preferred
Isolate specimens using press-to-seal silicone isolators with removable hydrophobic barriers. Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Two Sides Adhesive.
Thickness: 0.5 mm
Catalog No. 50-346-87
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.