Learn More
Neogen Diagnostics Red Stop Solution, 500 ML, Microwell
Supplier:  Neogen Diagnostics 301475
Encompass_Preferred
Red Stop Solution is a non-acidic, ready-to-use ELISA stop solution that contains no hazardous acids and can be used with our TMB substrates. Red Stop Solution does not increase background or optical density (OD) readings. Absorbance readings are taken at 650 nm within two hours of adding Red Stop Solution to the assay wells. Red Stop Solution offers a safer alternative to the traditional acids that are used to stop ELISA reactions. Stable for a minimum of 24-months at 2-8C or room temperature.
Catalog No. NC2215589
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.