Learn More
Safetec™ Red-Z™ Biohazard Fluid Control Powders
SDP
Catalog No. 19023899
Click to view available options
Type:
Green Z Spill Control Solidifier
Red Z Spill Control Solidifier
Packaging Quantity:
5 oz.
11 oz.
15 oz.
3.5 lb.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
19-023-899 Green Z Spill Control Solidifier 5 oz.
19-306-694 Red Z Spill Control Solidifier 11 oz.
19-023-901 Green Z Spill Control Solidifier 15 oz.
19-306-693 Red Z Spill Control Solidifier 5 oz.
19-290-179 Red Z Spill Control Solidifier 3.5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-023-899 Supplier Safetec™ Supplier No. 42010
Add to Cart
Add to Cart
For safe cleanup of spills of bodily fluids
Total fluid control encapsulators solidify and deodorize blood, vomit, urine or other potentially hazardous, water-based fluids. 
  • Red-Z solidifies and contains chlorine to deodorize potentially infectious blood and bodily fluids
  • Green-Z solidifies and can be used in the cleanup of blood, bodily fluids, chemotherapy, glutaraldehyde, and mercury spills
Order Info
Supplied in shaker-top bottles.
Specifications
Certifications/Compliance OSHA
Type Green Z Spill Control Solidifier
Format Powder
For Use With (Application) Incinerating, clean up of body fluids, chemical or mercury spills and recommended for use in hospitals, clinics, laboratories, nursing homes, schools and more
Packaging Quantity 5 oz.
Weight (Metric) 0.14 kg