Learn More
Safetec™ Red-Z™ Biohazard Fluid Control Powders

Description
Total fluid control encapsulators solidify and deodorize blood, vomit, urine or other potentially hazardous, water-based fluids.
- Red-Z solidifies and contains chlorine to deodorize potentially infectious blood and bodily fluids
- Green-Z solidifies and can be used in the cleanup of blood, bodily fluids, chemotherapy, glutaraldehyde, and mercury spills
Order Info
Supplied in shaker-top bottles.
Specifications
Specifications
| Certifications/Compliance | OSHA |
| Type | Red Z Spill Control Solidifier |
| Format | Powder |
| For Use With (Application) | Solidify and deodorize potentially Infectious blood and body fluids |
| Packaging Quantity | 3.5 lb. |
| Weight (Metric) | 1.5 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.