Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safetec™ Red-Z™ Biohazard Fluid Control Powders
SDP

Catalog No. 19306693
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Green Z Spill Control Solidifier
Red Z Spill Control Solidifier
Packaging Quantity:
5 oz.
11 oz.
15 oz.
3.5 lb.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
19-306-693 Red Z Spill Control Solidifier 5 oz.
19-306-694 Red Z Spill Control Solidifier 11 oz.
19-023-901 Green Z Spill Control Solidifier 15 oz.
19-290-179 Red Z Spill Control Solidifier 3.5 lb.
19-023-899 Green Z Spill Control Solidifier 5 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-306-693 Supplier Safetec™ Supplier No. 41101
Add to Cart
Add to Cart

For safe cleanup of spills of bodily fluids

Total fluid control encapsulators solidify and deodorize blood, vomit, urine or other potentially hazardous, water-based fluids. 

  • Red-Z solidifies and contains chlorine to deodorize potentially infectious blood and bodily fluids
  • Green-Z solidifies and can be used in the cleanup of blood, bodily fluids, chemotherapy, glutaraldehyde, and mercury spills

 

Order Info

Supplied in shaker-top bottles.

Specifications

Certifications/Compliance OSHA
Type Red Z Spill Control Solidifier
Format Powder
For Use With (Application) Solidify and deodorize potentially Infectious blood and body fluids
Packaging Quantity 5 oz.
Weight (Metric) 0.14 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.