Learn More
Abcam Reduced Cytochrome C, 1 mL
Description
- Product: Reduced Cytochrome C
- Cited in: > 2 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | Unconjugated |
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Cytochrome C |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.