Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REEP2 Monoclonal Antibody (OTI2B7)
GREENER_CHOICE

Catalog No. PIMA525878
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525878 100 μL
1 options

Catalog No. PIMA525878

Supplier: Invitrogen™ MA525878

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

REEP2 Monoclonal Antibody for Western Blot, Flow

Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2B7
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene REEP2
Gene Accession No. Q8VCD6, Q9BRK0
Gene Alias C2orf19; C5orf19; LOC532432 protein; LOC682105; receptor accessory protein 2; receptor expression-enhancing protein 2; Reep2; SGC32445; SPG72; Yip2d
Gene Symbols REEP2
Host Species Mouse
Immunogen Full length human recombinant protein of REEP2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100683708, 225362, 51308, 682105
Target Species Human, Mouse, Rat, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.