Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reference Color Standard No. 9, ASTM D 848, Ricca Chemical
SDP

Catalog No. R6605900500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6605900500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R6605900500 Supplier Ricca Chemical Company Supplier No. R6605900500C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-50-9 , 7789-00-6 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Packaging Amber Glass Bottle
Physical Form Liquid
Color Yellow
For Use With (Equipment) Water Analyzer
Solubility Infinitely soluble in water
Solution Type Calibration Solution
Type Color Reference Standard
Solvent or Matrix Water
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Reference Color Standard No. 9, ASTM D 848
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.