Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Refill for HAND MEDIC™ ADX-7™ Dispenser
SDP

Catalog No. 19043855 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
685 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-855 685 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-855 Supplier GOJO™ Supplier No. 874504
  • $153.00
    $110.65 / Case of 4

    Save $42.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for trouble-free performance with engineering grade plastics construction. Dispenser features a compact design, making it ideal for tight spaces.

  • Fragrance-free skin conditioner
  • Specially formulated for industrial environments
  • Helps prevent dry, cracked skin by maintaining skin's moisture level
  • Absorbs quickly with no greasy after-feel
  • Silicone and fragrance free
  • Dermatologist tested
  • Latex glove compatible
  • NSF E4 Registered
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with each refill
  • SMART-FLEX™ bottle design uses 30% less material than HDPE
  • Patent-pending controlled collapse technology maintains bottle shape longer while emptying
  • Removable pump for easier recycling
TRUSTED_SUSTAINABILITY

Specifications

Product Type Skin Conditioner
Color White
Form Lotion
For Use With (Application) Helps prevent dry, cracked skin by maintaining skin moisture level
For Use With (Equipment) 8781-06, or 8782-06
Quantity 4/Pk.
Size 685 mL
Volume (Metric) 685 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.