Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Reflection & Refraction Tank
SDPSCI_ED

Catalog No. S43683 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43683 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43683 Supplier United Scientific Supplies Supplier No. RRT002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product demonstrates the laws of reflection and refraction.

  • This product demonstrates the laws of reflection and refraction.
  • The 16 cm diameter acrylic water tank has a 360° scale and a moveable arm with a push-button activated laser.
  • Thumb screws on the side of the laser harness can be adjusted to finely position the laser beam.
  • Requires two AAA batteries (not included).
  • Includes activity guide.

Specifications

Grade 5 to 12
Format Kit
Product Type Reflection Demonstrator
Languages English
Subject Physics - Optics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.