Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Peroxide Test Strips

Catalog No. M1094500013 Shop All MilliporeSigma Products
Change view
Click to view available options
Range:
0.2 to 20mg/L
100 to 1000mg/L
pH 2.0 to 9.0
Type:
Test Strip
pH Strip
No. of Tests:
50
25000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Type No. of Tests
M1094500013 pH 2.0 to 9.0 pH Strip 25000
M1169740001 0.2 to 20mg/L Test Strip 50
M1167310001 100 to 1000mg/L Test Strip 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1094500013 Supplier MilliporeSigma™ Supplier No. 1.09450.0013

Includes

Tube containing 50 strips, 1 bar-code strip.

For determination of inorganic peroxides in aqueous solutions

  • Enzymatic reaction method ideal for screening
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

TRUSTED_SUSTAINABILITY

Specifications

pH Range 2 to 9
For Use With (Application) For food and water analysis
Graduations 2-2.5-3-3.5-4-4.5-5-5.5-6-6.5-7-7.5-8-8.5-9
No. of Tests 25000
Range pH 2.0 to 9.0
Storage Requirements 2°C to 8°C
Type pH Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.