Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Scientific Reflex Hammer
SCI_ED

Catalog No. S02882 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
Bones and Muscles Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S02882 Bones and Muscles Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02882 Supplier AquaPhoenix Scientific Supplier No. HA5075R
Add to Cart
Add to Cart

Durable reflex hammer featuring a triangular rubber head—perfect for demonstrating deep tendon reflexes in anatomy, physiology, or medical training settings.

  • Classic triangular rubber head for accurate reflex testing
  • Used to elicit deep tendon reflexes (DTRs)
  • Ideal for anatomy, neurology, and clinical skills demonstrations
  • Sturdy handle for controlled tapping
  • Lightweight and easy to handle
  • For educational and demonstration purposes only
  • Commonly used in biology, pre-med, and health science classrooms

Specifications

Grade 9 to 12
Class Size 1 Lab Group
Product Type Bones and Muscles Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.