Learn More
Science First™ Refracting Telescope Kit
SDPSCI_ED
Catalog No. S48823 Shop All Science First Products
Change view
Click to view available options
Quantity:
10 kits
Single Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S48823 10 kits
S48822 Single Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S48823 Supplier Science First™ Supplier No. 6540010
Add to Cart
Add to Cart
Includes
All components are included: plastic lenses, coated cardboard telescope tubes, and instructions.
Students make a working refracting telescope
This Refracting Telescope Kit allows students to repeat Galileo's ground-breaking work, by constructing 16X refracting telescope. Along the way, students will be able to identify the elements of a simple refracting telescope, learn how lenses work, and be able to measure telescope magnification. No expensive optical benches or materials are required.
Specifications
For Use With (Application) Study how lenses work when put together without using an optical bench
Includes Inner and outer tubes, plastic eyepiece lenses, foam holders, cardboard spacers and washers for the eyepiece lenses, red plastic caps and 1 set of instructions and activitie
Quantity 10 kits
Length (Metric) 400 mm and 25 mm focal length
Diameter (Metric) Outer 43 mm and 17.5 mm Lens Diameter
Product Type Refracting telescope Kit