Learn More
Thermo Scientific™ Refrigerator and Freezer Door Options
Catalog No. 12370628
Change view
Click to view available options
For Use With (Equipment):
11.5 cu. ft. Units
23.3 cu. ft. Units
29.2 cu. ft. Units
4.7 cu. ft. Forma, Jewett, Puffer Hubbard
4.7 cu. ft. Units
45.8 cu. ft. Units
Type:
Refrigerator and Freezer Door, Glass
Refrigerator and Freezer Door, Heated Glass
Refrigerator and Freezer Door, Solid
Hinge Side:
Left
Left/Left
Left/Right
Right
Right/Right
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type Hinge Side
12-370-628 23.3 cu. ft. Units Refrigerator and Freezer Door, Solid Left
12-370-627 11.5 cu. ft. Units Refrigerator and Freezer Door, Glass Left
12-370-630 29.2 cu. ft. Units Refrigerator and Freezer Door, Solid Left
12-370-632 45.8 cu. ft. Units Refrigerator and Freezer Door, Solid Left/Left
12-370-633 23.3 cu. ft. Units Refrigerator and Freezer Door, Glass Right
12-370-637 45.8 cu. ft. Units Refrigerator and Freezer Door, Glass Left/Left
12-370-639 45.8 cu. ft. Units Refrigerator and Freezer Door, Glass Right/Right
12-370-640 11.5 cu. ft. Units Refrigerator and Freezer Door, Heated Glass Left
12-370-642 23.3 cu. ft. Units Refrigerator and Freezer Door, Heated Glass Left
12-370-644 29.2 cu. ft. Units Refrigerator and Freezer Door, Heated Glass Left
12-370-645 29.2 cu. ft. Units Refrigerator and Freezer Door, Heated Glass Right
12-370-646 45.8 cu. ft. Units Refrigerator and Freezer Door, Heated Glass Left/Left
12-370-648 45.8 cu. ft. Units Refrigerator and Freezer Door, Heated Glass Left/Right
12-370-652 4.7 cu. ft. Units Refrigerator and Freezer Door, Glass Left
12-370-629 23.3 cu. ft. Units Refrigerator and Freezer Door, Glass Left
12-370-651 4.7 cu. ft. Forma, Jewett, Puffer Hubbard Refrigerator and Freezer Door, Solid Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Door options must be specified at time of order.
Catalog No. 12-370-628 Supplier Thermo Scientific™ Supplier No. 6997
Add to Cart
Add to Cart
Door options must be specified at time of order.
Customize refrigerator and freezer configurations to better suit the needs of the lab with these refrigerator and freezer door options.
Customize refrigerator and freezer configurations to better suit the needs of the lab with Thermo Scientific™ Refrigerator and Freezer Door Options which include glass and heated glass doors, as well as either right- or left-hand swing.
For use with Thermo Scientific Forma, Jewett, Puffer Hubbard, and Revco series. Solid doors are standard on most refrigerators unless otherwise specified. Optional dual-pane glass doors must be specified when ordering. Contact your Sales Representative for information about heated glass doors.
  • Glass doors standard on selected blood bank, chromatography and pharmacy refrigerators
  • Sliding glass doors standard on all 45.8 cu. ft. (1297L) models
  • Heated glass doors increase amperage and may require next size breaker
Ordering Information:
Door options must be specified at time of order
TRUSTED_SUSTAINABILITY
Specifications
Type Refrigerator and Freezer Door, Solid
Capacity (English) 45.8 cu. ft.
Capacity (Metric) 1297 L
Door Style Hinged
Door Material Glass (Door)
Hinge Side Left
For Use With (Equipment) 23.3 cu. ft. Units
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.