Learn More
Darwin Chambers Company Single Door Lab Refrigerator 21 cu. ft.
SDP
Supplier:  Darwin Chambers Company LT030AA
Encompass_Preferred
The LT030 is a 23.9 cubic feet single door laboratory refrigerator. Temperature range is 2°C to 15°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. Our LT series laboratory refrigerators provide the highest level of performance and construction with tight uniformity and control of temperature.
Catalog No. 50-189-3912
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.