Learn More
Darwin Chambers Company Double Door Lab Refrigerator 47 cu. ft.
SDP
Supplier:  Darwin Chambers Company LT055SS
Encompass_Preferred
The LT055-SS is a 50 cubic feet double door laboratory refrigerator. Temperature range is 2°C to 15°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Our LT series laboratory refrigerators provide the highest level of performance and construction with tight uniformity and control of temperature.
Catalog No. 50-189-3915
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.