Learn More
Caron Products And Services Refrigerated Incubator, 2-Door, 50 cu. ft., Temp, No RH
Supplier: Caron Products And Services 7001502
50 ft3 (1,416 L) Refrigerated Incubator offers precise temperature control (5C to 70C). Ideal for a wide range of applications. This reach-in unit delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.