Learn More
Invitrogen™ REG4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total REG4 protein.
Specifications
Specifications
| Antigen | REG4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | REG4 |
| Gene Accession No. | Q9BYZ8 |
| Gene Alias | 2010002L15Rik; Gastrointestinal secretory protein; GISP; GISPREG-4; Reg IV; REG4; REG-4; regenerating family member 4; regenerating gene type IV; regenerating islet-derived family, member 4; regenerating islet-derived protein 4; Regenerating islet-derived protein IV; REG-IV; REG-like protein; RELP |
| Gene Symbols | REG4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.