Learn More
Cytiva Regeneration Scouting Kit, 1kit
Catalog No. 501055332 Shop All Cytiva Products
Encompass
Contains 10 solutions, most ready-to-use, for developing regeneration conditions for interaction analyses in Biacore systems.
Catalog No. 50-105-5332 Supplier Cytiva Supplier No. BR100556
Contains 10 solutions, most ready-to-use, for developing regeneration conditions for interaction analyses in Biacore systems.