Learn More
Regis Technologies Inc GC Regeant, Regis, REGISIL (BSTFA) + 10% TMCS (25 GM)
SDP
Supplier:  Regis Technologies Inc 1270133200
Encompass_Preferred
BSTFA +10% TMCS-Regisil is a Silylation Reagent. BSTFA reacts faster and more completely than BSA, due to the trifluoroacetamide leaving group. The high volatility of BSFTA and its byproducts results in non-co-elution of early eluting peaks. Additionally, the highly volatile and stable products result in low detector noise and fouling. Addition of TMCS to BSTFA catalyzes reactions of hindered functional groups and other difficult functionalities, such as secondary hydroxyls and amines.
Catalog No. 50-442-888
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.