Learn More
Vikan™ Regular Undercarriage
Catalog No. 17227655 Shop All Remco Products
Change view
Click to view available options
Color:
Steel
Catalog No. Color
17-227-655 Steel
1 options
Catalog No. 17-227-655 Supplier Vikan™ Supplier No. 6903
Add to Cart
Add to Cart
Regular Undercarriage is commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications.
  • Tall
  • Galvanized steel undercarriage
  • Features caster wheels that are impervious to meat fat and bearings equipped with grease fittings
  • Includes a built-in stop that allows the tub to be tipped and emptied
  • Color-coded
Specifications
Product Type Undercarriage
Color Steel
Material Galvanized Steel
Quantity 1
Dimensions (L x W x H) 41.1 x 27.3 x 19.1 in.