Learn More
Medchemexpress LLC Rehmannioside D | 81720-08-3 | MFCD30478904 | 99.9% | 686.61 g/mol | C27H42O20 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN091210MG
Encompass_Preferred
Rehmannioside D is a carotenoid glycoside isolated from the roots of Rehmannia glutinosa. Supplied as a 10 mg powder research reagent, it is used as an analytical standard and in biochemical studies of antioxidant activity and reactive oxygen species (ROS). Molecular formula C27H42O20; molecular weight 686.61 g/mol.
  • Purity 99.9% suitable for analytical applications.
  • Powder form, stable under recommended storage conditions.
  • Useful in antioxidant and ROS pathway studies.
  • Provided as a 10 MG vial for small-scale experiments.
  • Identified by CAS number 81720-08-3 for unambiguous chemical identification.
Catalog No. 50-002-96213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.