Learn More
Reichert™ Sample Press for AR70 and AR700 Refractometers
Catalog No. 13975122 Shop All Reichert, Inc Products
Change view
Click to view available options
Sample Press
Catalog No.
13-975-122 Sample Press
1 options
Catalog No. 13-975-122 Supplier Reichert™ Supplier No. 13107300
Accessory for use with AR70 and AR700 Refractometers.
  • For AR7 series refractometers
  • Ideally suited for thick, viscous sample types
  • Press forms a uniform contact bond of the sample with the prism-measuring surface
  • Sample press provides an enclosed environment that is helpful for sample types that tend to evaporate, which would change the refractive index and concentration of the sample
Specifications
Type Sample Press
For Use With (Equipment) AR7 Series Refractometers AR70 and AR700