Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reinecke Salt, ACS Reagent, ≥93.0%, Solstice

Catalog No. 60045031 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-031 25 g Glass Bottle
60-045-030 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-031 Supplier Solstice Supplier No. 18368725G

missing translation for 'validMainframeMsg'

ACS reagent,≥93.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-16-5
MDL Number MFCD00066638

Specifications

Melting Point 272°C
Physical Form Solid
Quantity 25 g
Linear Formula NH4[Cr(NH3)2(SCN)4]
UN Number NONH for all modes of transport
Molecular Weight (g/mol) 336.43
Formula Weight 336.43g/mol
Percent Purity ≥93.0%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Reinecke Salt
Show More Show Less
Hazard Statement P261-P280-P301 + P312 + P330
Precautionary Statement H302 + H312 + H332
EINECSNumber 237-003-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.