Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Relaxin-2 Antibody (349121) [CoraFluor™ 1], R&D Systems™
SDP

Catalog No. FAB28041CL1
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB28041CL1 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB28041CL1 Supplier R&D Systems Supplier No. FAB28041CL1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

Relaxin-2 Monoclonal antibody specifically detects Relaxin-2 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry

Specifications

Antigen Relaxin-2
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 349121
Conjugate CoraFluor 1
Formulation PBS
Gene Alias bA12D24.1.1, bA12D24.1.2, H2, relaxin 2, RLXH2
Host Species Rat
Immunogen E. coli-derived recombinant human Relaxin-2, Val23-Cys185, Accession # P04090
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6019
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.