Learn More
Medchemexpress LLC HY-16752 100mg , Relebactam CAS:1174018-99-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16752100MG
Encompass_Preferred
HY-16752 100mg Relebactam CAS: 1174018-99-5 Relebactam is a diazabicyclooctane inhibitor with activity against a wide spectrum of β-lactamases, including class A (extended-spectrum β-lactamases and KPC) and class C (AmpC) enzymes. Relebactam shows antibacterial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9771