Learn More
Description
- Automated DNA Isolation from 1 to 10mL Blood
- Effective recovery from pristine, hemolysed or frozen samples
- Purified DNA ready for use in downstream assays or archiving
- Chemistry scaled for each sample, reducing waste
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Product Type | gDNA Isolation Kit |
| Quantity | 1/Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.