Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523910
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523910 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523910 Supplier Thermo Scientific Supplier No. PA523910
Only null left
Add to Cart
Add to Cart

RELL2 Polyclonal Antibody

RELL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RELL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RELL2
Gene Accession No. Q8BRJ3, Q8NC24
Gene Alias 4631403P03Rik; C5orf16; ependolin; receptor expressed in lymphoid tissues like 2; Rell2; RELT like 2; RELT-like 2; RELT-like protein 2; UNQ9423/PRO34565
Gene Symbols RELL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 132-160 amino acids from the central region of human RELL2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225392, 285613
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.