Learn More
Invitrogen™ RELM alpha Monoclonal Antibody (DS8RELM), APC, eBioscience™

Description
This DS8RELM monoclonal antibody recognizes mouse RELM alpha, also known as FIZZ1. This antibody was shown not to cross-react with mouse RELM gamma. Applications Reported: This DS8RELM antibody has been reported for use in flow cytometric analysis. Applications Tested: This DS8RELM antibody has been tested by flow cytometric analysis of resident peritoneal macrophages using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Specifications
Specifications
| Antigen | RELM alpha |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DS8RELM |
| Concentration | 0.2 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Retnla |
| Gene Accession No. | Q9EP95 |
| Gene Alias | 1810019L16Rik; cysteine-rich secreted protein A12-gamma; Cysteine-rich secreted protein FIZZ1; Fizz1; Fizz-1; FIZZ2; found in inflammatory zone 1; Himf; HXCP2; Hypoxia-induced mitogenic factor; parasite-induced macrophage novel gene 1 protein; Pmng1; RELMa; RELMalpha; RELM-alpha; RELMb; RELMbeta; RELM-beta; resistin like alpha; resistin-like alpha; Retnla; Xcp2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.