Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Bordetella parapertussis

Catalog No. R30165501
Change view
Click to view available options
Description:
Bordetella Parapertussis Sera
Bordetella Pertusis Sera
Quantity:
2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
R30165501 Bordetella Pertusis Sera 2 mL
R30165601 Bordetella Parapertussis Sera 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R30165501 Supplier Thermo Scientific™ Supplier No. R30165501
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for serological identification of Bordetella cultures by rapid slide agglutination test.

Remel Agglutinating Sera are used for serological identification of Bordetella cultures by rapid slide agglutination test.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Bordetella Pertusis Sera
CE Marker Yes
Format Vial
Packaging Type 2 mL/Vial
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.