Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Catalog No. R30954901
Change view
Click to view available options
Description:
E. coli Polyvalent 2
E. coli Polyvalent 3
E. coli Polyvalent 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30954901 E. coli Polyvalent 2
R30955101 E. coli Polyvalent 4
R30955001 E. coli Polyvalent 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R30954901 Supplier Thermo Scientific™ Supplier No. R30954901
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
TRUSTED_SUSTAINABILITY
Specifications
Description E. coli Polyvalent 2
Detectable Analytes E. coli
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Specificity types O26, O55, O111, O119, O126