Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Description
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Specifications
Specifications
| Description | Group W135 |
| CE Marker | Yes |
| Detectable Analytes | N. meningitidis |
| Type | Neisseria Meningitidis Test Kit |
| Organism Group | Group W135 |
| Target Organism Class | Neisseria Meningitidis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.