Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Specifications
| Control Sets | Positive |
| Description | Salmonella H (a) |
| CE Marker | Yes |
| Detectable Analytes | Salmonella H Antigen |
| Type | Agglutinating Sera |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |
| Target Organism Class | Salmonella |