Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Specifications
Control Sets Positive
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Material Polyvalent
Packaging Type 2 mL/Vial
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
R30858101
View Documents
Thermo Scientific™
R30858101
Polyvalent (group A - G)
Each for $618.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
R30858201
View Documents
Thermo Scientific™
R30858201
Polyvalent (group A - S)
Each for $939.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Specifications
Positive
Agglutinating Sera
2 mL/Vial
Salmonella O
Salmonella O Antigen
Polyvalent
2 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.