Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O

Catalog No. R30956701
Change view
Click to view available options
Description:
Factor 11 (group F)
Factor 2 (group A)
Factor 4 (group B )
Factor 5 (group B)
Factor 6, 7 (group C1)
Factor 8 (group C2)
Factor 9 (group D)
Factors 3, 10, 15, 19 (group E)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30956701 Factor 2 (group A)
R30956801 Factors 3, 10, 15, 19 (group E)
R30957601 Factor 11 (group F)
R30957101 Factor 6, 7 (group C1)
R30957201 Factor 8 (group C2)
R30957001 Factor 5 (group B)
R30957301 Factor 9 (group D)
R30956901 Factor 4 (group B )
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. R30956701 Supplier Thermo Scientific™ Supplier No. R30956701
Add to Cart
Add to Cart
This item is not returnable. View return policy

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Factor 2 (group A)
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Organism Group Group A
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.