Learn More
Thermo Scientific™ APT Broth
Catalog No. R452312 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452312 500 g
1 options
This item is not returnable. View return policy
Catalog No. R452312 Supplier Thermo Scientific™ Supplier No. R452312
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cultivate lactic acid bacteria and other organisms that require high thiamine content with this liquid medium.
Cultivate lactic acid bacteria and other organisms that require high thiamine content with Thermo Scientific™ Remel™ APT Broth.
Formulated by Evans and Niven1 to cultivate the heterofermentative lactobacilli that causes green discoloration of cured meat products. Deibel used APT Broth for isolation of bacteria that requires thiamine such as Lactobacillus viridescens2. The American Public Health Association (APHA)3 mentions APT Broth that enumerates and cultivates lactic acid bacteria associated with bacterial spoilage of dairy products, meats and vegetables.
Cultivate heterofermentative lactic acid bacteria using the liquid medium APT Broth.
  • Easy-to-cultivate and identify: lactic acid bacteria such as Streptococcus, Leuconostoc, Lactobacillus and Pediococcus associated with bacterial spoilage of dairy products, meats and vegetables.
  • Recommended by the American Public Health Association (APHA) for enumeration and cultivation of lactic acid bacteria associated with bacterial spoilage of dairy products, meat and vegetables.
APT Broth contains Casein peptone, yeast extract, sodium chloride, dextrose, metallic salts ions, polysorbate 80 and agar that provides nitrogen, amino acids, peptides necessary for bacterial growth, provides B-complex that serves as a growth enhancer, maintains osmotic equilibrium, acts as a ready source of energy, essential for replication of lactic acid bacteria, acts as a source of fatty acids for lactobacilli, and acts as a solidifying agent, respectively.
APT Broth is nonselective. It favors growth of many other bacteria other than Lactobacillus4.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description APT Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only