Learn More
Thermo Scientific™ Remel™ Baird-Parker Agar Base
Catalog No. R452342 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452342 500 g
1 options
This item is not returnable. View return policy
Catalog No. R452342 Supplier Thermo Scientific™ Supplier No. R452342
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and presumptively identify coagulase positive Staphylococci from foods and other sources using Baird-Parker Agar Base (Dehydrated).
Isolate and enumerate coagulase positive Staphylococci from foods and other sources1 using Thermo Scientific™ Remel™ Baird-Parker Agar Base (Dehydrated). Zebovitz, Evan, and Nivan developed Baird-Parker medium by modifying tellurite glycine medium2. This medium is recommended for use in the examination of foods and other materials by the Food and Drug Administration (FDA), the American Public Health Association (APHA), and the United States Pharmacopeia (USP)3-7.
Use Baird-Parker Agar Base for isolation and enumeration of coagulase positive Staphylococci from foods and other sources.
  • Selective solid medium - Lithium chloride acts as a selective agent which inhibits most bacteria other than S. aureus.
  • Easy-to-read: Staphylococcus aureusforms grey-black shiny colonies with surrounding clear zones.
  • Recommended for use in the examination of foods and other materials by the Food and Drug Administration (FDA), the American Public Health Association (APHA), and the United States Pharmacopeia (USP)3-7.
This medium contains beef extract, peptone, and yeast extract which supplies nitrogen, carbon, sulfur, vitamins, and trace minerals. Sodium pyruvate and glycine enhance the growth of S. aureus. S. aureus reduces potassium tellurite to metallic tellurium resulting in black colonies. On further incubation many strains of S. aureus form opaque zones within the clear zones because of lecithinase or lipase activity.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, EP, FDA, IDF, ISO, NMKL, USDA.
Description Baird-Parker Agar Base
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only