Learn More
Thermo Scientific™ Brain Heart Infusion (BHI) Agar
Description
Isolate and cultivate a wide variety of micro-organisms including pathogenic fungi this ready-to-use medium recommended by AOAC and APHA.
Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar. Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2
Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.
- Recommended by AOAC and APHA.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, COMPF, EPA, IDF, SMD. |
| Description | Brain Heart Infusion Agar (BHI) |
| Format | 20mm x 113mm Tube - Slant |
| Manufacturing Origin | United States |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |
REF R112025 not intended for IVD use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.