Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brain Heart Infusion (BHI) Agar

Catalog No. R09060
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
20mm x 113mm Tube - Slant
20mm x 150mm Pour Tube
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R09060 20/Pk. 20mm x 150mm Pour Tube
R060232 20/Pk. 15mm x 103mm Tube
R08060 100/Pk. 20mm x 113mm Tube - Slant
R08062 20/Pk. 20mm x 113mm Tube - Slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R09060 Supplier Thermo Scientific™ Supplier No. R09060
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and cultivate a wide variety of micro-organisms including pathogenic fungi this ready-to-use medium recommended by AOAC and APHA.

Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar. Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2

Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.

  • Recommended by AOAC and APHA.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, EPA, IDF, SMD.
Description Brain Heart Infusion Agar (BHI)
Format 20mm x 150mm Pour Tube
Manufacturing Origin United States
Product Type Agar Plate
Quantity 20/Pk.

REF R112025 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.