Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin
Description
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
- Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
- Gentamicin inhibits some gram-negative bacteria
Specifications
Specifications
| Description | BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin |
| Format | F - slant |
| Product Type | Agar |
| Quantity | 20/Pk. |