Learn More
Description
Selectively isolate and presumptively identify group D Streptococci and Enterococci using Bile Esculin Azide Agar (Dehydrated).
Selectively isolate and presumptively identify group D Streptococci and Enterococci using Thermo Scientific™ Remel™ Bile Esculin Azide Agar (Dehydrated). Esculin agar containing 40% bile, correlated well with serologically confirmed group D streptococci as reported by Swan1. Isenberg et al. modified the formulation by reducing the concentration of bile and adding the selective agent, sodium azide2.
In the presence of bile, group D Streptococci and Enterococci hydrolyze esculin and form esculetin and dextrose. Ferric ammonium citrate supplies ferric ions which react with esculetin and form brown-black colonies on Bile Esculin Agar.
- Selective solid media - Selective agents like sodium azide and 1% Oxgall inhibit Bacilli and most gram-positive bacteria other than group D Streptococci and Enterococci.
- Easy-to-read – black-brown colonies of group D Streptococci and Enterococci are easy to read.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, SMWW. |
| Description | Bile Esculin Azide Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.