Learn More
Thermo Scientific™ Remel™ Biplates: Anaerobic Reducible through Levine Agars
GSA_VA
Catalog No. R02058
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Media Type:
Chocolate /Thayer Martin Agar
Columbia CNA/MacConkey Agar
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
R02058 10/Pk. Chocolate /Thayer Martin Agar
R02065 10/Pk. Columbia CNA/MacConkey Agar
R02066 100/Pk. Columbia CNA/MacConkey Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R02058 Supplier Thermo Scientific™ Supplier No. R02058
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel™ Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Format Biplate
Media Type Chocolate /Thayer Martin Agar
Product Type Agar Plate
Quantity 10/Pk.