Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Birdseed Agar

Catalog No. R09176
Change view
Click to view available options
Format:
20mm x 150mm Pour Tube
Monoplate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R09176 20mm x 150mm Pour Tube
R01192 Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R09176 Supplier Thermo Scientific™ Supplier No. R09176

Thermo Scientific™ Remel Birdseed Agar is for the presumptive identification of Cryptococcus neoformans.

Thermo Scientific™ Remel Birdseed Agar is for the presumptive identification of Cryptococcus neoformans.

TRUSTED_SUSTAINABILITY

Specifications

Description Birdseed Agar
Format 20mm x 150mm Pour Tube
Product Type Agar Tube
Quantity 20 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.