Learn More
Thermo Scientific™ Remel™ Bordet Gengou Agar Base
Catalog No. R452432 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452432 500 g
1 options
This item is not returnable. View return policy
Catalog No. R452432 Supplier Thermo Scientific™ Supplier No. R452432
Add to Cart
Add to Cart
This item is not returnable. View return policy
Remel Bordet Gengou Agar Base is used for the isolation of Bordetella pertussis and Bordetella parapertussis.
Remel Bordet Gengou Agar Base is used for the isolation of Bordetella pertussis and Bordetella parapertussis.
TRUSTED_SUSTAINABILITY
Specifications
Description Bordet Gengou Agar Base
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only