Learn More
Thermo Scientific™ Brilliant Green Agar with Novobiocin
Description
Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with this selective medium.
Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with Thermo Scientific™ Remel™ Brilliant Green Agar with Novobiocin. The medium is recommended by the American Public Health (APHA) for the recovery of Salmonella from food products1. Moats added novobiocin to the medium to inhibit growth of Proteus in culture from beef and poultry products2.
Specifications
Specifications
| Description | Brilliant Green Agar w/Novobiocin |
| Format | Monoplate |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |
REF R110113 not intended for IVD use.