Learn More
Thermo Scientific™ Remel™ Brucella Broth (Dehydrated)
Catalog No. R452662 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452662 500 g
1 options
Catalog No. R452662 Supplier Thermo Scientific™ Supplier No. R452662
Add to Cart
Add to Cart
Cultivate Brucella spp. and other fastidious microorganisms using Brucella Broth (Dehydrated).
Cultivate Brucella species and a variety of microorganisms using Thermo Scientific™ Remel™ Brucella Broth (Dehydrated). Brucella Broth prepared according to the formula of Albimi Broth is recommended by the American Public Health Association1. Albimi Broth is used to isolate Brucella from dairy products, and foods2. Brucella Broth is used for preliminary enrichment of Brucella and it is isolated using Brucella Broth in a biphasic medium.
This medium contains casein and meat peptones which provide nitrogen, amino acids and peptides necessary for bacterial growth. Sodium bisulfite is a reducing agent. Yeast extract is a source of B-complex vitamins. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. Dextrose provides a ready source of energy.
  • Easy to prepare - Convenient to use
  • Easy to store - No special storage required
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: USDA.
Description Brucella Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g