Learn More
Thermo Scientific™ Remel™ Buffered CYE Agar Base
Catalog No. R452352 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452352 500 g
1 options
This item is not returnable. View return policy
Catalog No. R452352 Supplier Thermo Scientific™ Supplier No. R452352
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Remel Buffered CYE Agar Base is used for the isolation of Legionella spp.
Remel Buffered CYE Agar Base is used for the isolation of Legionella spp.
Also available: Buffered CYE Supplement, Part No R450041; PAC Supplement, Part No. R45006; and PAV Supplement, Part No. R45007.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffered CYE Agar Base
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only