Learn More
Thermo Scientific™ Remel™ Buffered CYE Agar
Catalog No. R07092
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
16mm x 125mm Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R07092 16mm x 125mm Tube
R01342 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R07092 Supplier Thermo Scientific™ Supplier No. R07092
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use for the cultivation and isolation of Legionella species.
Thermo Scientific™ Remel Buffered CYE Agar (Buffered Charcoal Yeast Extract) (BCYE) is used for the cultivation and isolation of Legionella species.
Ordering Information:
REF R09300 requires addition of BCYE supplement, REF R45005. Optional antimicrobial supplements are PAC, REF R45006, PAV, REF R45007.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffered CYE Agar (BCYE)
Format 16mm x 125mm Tube
Product Type Agar Plate
Quantity 20/Pk.