Learn More
Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer
Description
Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, USDA, USP. |
| Description | Phosphate Buffer, Butterfield's |
| Format | Glass Vial - 9mL |
| Product Type | Buffer Solution |
| Packaging Type | 100/Pk. |
| Quantity | 100 x 9 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.